Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGTTCTTCCTTCCATGGCAGGGG[C/T]TACGTGGGCACCGGGCGCTTGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602378 MIM: 605395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf38 - chromosome 19 open reading frame 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNM2 - dynamin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6793 - microRNA 6793 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED1 - transmembrane p24 trafficking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006858.3 | 800 | Silent Mutation | TAA,TAG | *,* 228 | NP_006849.1 | |
XM_006722631.3 | 800 | Silent Mutation | TAA,TAG | *,* 190 | XP_006722694.1 |