Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTGAACCATATCTGAAGGTTT[C/T]CCCACACTGCTTACACACATAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF573 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF573 - zinc finger protein 573 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172689.1 | 1869 | Missense Mutation | AAA,GAA | K,E 557 | NP_001166160.1 | |
NM_001172690.1 | 1869 | Missense Mutation | AAA,GAA | K,E 645 | NP_001166161.1 | |
NM_001172691.1 | 1869 | Missense Mutation | AAA,GAA | K,E 643 | NP_001166162.1 | |
NM_001172692.1 | 1869 | Missense Mutation | AAA,GAA | K,E 557 | NP_001166163.1 | |
NM_152360.3 | 1869 | Missense Mutation | AAA,GAA | K,E 587 | NP_689573.3 | |
XM_011526446.2 | 1869 | Missense Mutation | AAA,GAA | K,E 660 | XP_011524748.1 | |
XM_017026275.1 | 1869 | Missense Mutation | AAA,GAA | K,E 660 | XP_016881764.1 | |
XM_017026276.1 | 1869 | Missense Mutation | AAA,GAA | K,E 660 | XP_016881765.1 | |
XM_017026277.1 | 1869 | Missense Mutation | AAA,GAA | K,E 658 | XP_016881766.1 | |
XM_017026278.1 | 1869 | Missense Mutation | AAA,GAA | K,E 589 | XP_016881767.1 | |
XM_017026279.1 | 1869 | Missense Mutation | AAA,GAA | K,E 589 | XP_016881768.1 | |
XM_017026280.1 | 1869 | Missense Mutation | AAA,GAA | K,E 557 | XP_016881769.1 | |
XM_017026281.1 | 1869 | Missense Mutation | AAA,GAA | K,E 557 | XP_016881770.1 |