Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCAACTTCCATCAGTGGCGCGA[C/T]GCTCGCCAGGTCTGGGGCCTCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606580 MIM: 601703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPA3 - optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VASP - vasodilator-stimulated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003370.3 | 582 | Silent Mutation | GAC,GAT | D,D 84 | NP_003361.1 | |
XM_005259199.1 | 582 | Silent Mutation | GAC,GAT | D,D 84 | XP_005259256.1 | |
XM_005259200.1 | 582 | Silent Mutation | GAC,GAT | D,D 84 | XP_005259257.1 | |
XM_017027200.1 | 582 | Silent Mutation | GAC,GAT | D,D 84 | XP_016882689.1 |