Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTTTGAGTCCCGGAATCTGAGA[C/T]GGTGTGCACCCTTCCCCAGCTCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602649 MIM: 602756 | ||||||||||||||||||||
Literature Links: |
C19orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C19orf24 - chromosome 19 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIRBP - cold inducible RNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIRBP-AS1 - CIRBP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA2 - ephrin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |