Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCATGGGGAACTTCAGGTGGGC[A/G]CACCTCAAACATGATGGTGATGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607430 MIM: 601240 MIM: 601825 | ||||||||||||||||||||
Literature Links: |
DAZAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAZAP1 - DAZ associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAMT - guanidinoacetate N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000156.5 | 668 | Intron | NP_000147.1 | |||
NM_138924.2 | 668 | Missense Mutation | CGC,TGC | R,C 192 | NP_620279.1 |
NDUFS7 - NADH:ubiquinone oxidoreductase core subunit S7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |