Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAATGTGATGTTGGAGAACTTTG[C/T]GCTTTTGTCATCAGTAGGTAAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF749 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF749 - zinc finger protein 749 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023561.3 | 1379 | Missense Mutation | GCG,GTG | A,V 42 | NP_001018855.2 | |
NM_001321952.1 | 1379 | Missense Mutation | GCG,GTG | A,V 35 | NP_001308881.1 | |
NM_001321953.1 | 1379 | Intron | NP_001308882.1 | |||
NM_001321954.1 | 1379 | Intron | NP_001308883.1 | |||
XM_011526956.2 | 1379 | Missense Mutation | GCG,GTG | A,V 35 | XP_011525258.1 | |
XM_017026802.1 | 1379 | Missense Mutation | GCG,GTG | A,V 42 | XP_016882291.1 |