Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACGACAAGGCAGGAACAGAAAG[C/T]GAGCAGGGCAGCGACGCCAGCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIGLEC11 - sialic acid binding Ig like lectin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135163.1 | 1704 | Intron | NP_001128635.1 | |||
NM_052884.2 | 1704 | Intron | NP_443116.2 | |||
XM_005258476.3 | 1704 | Missense Mutation | ACT,GCT | T,A 558 | XP_005258533.2 | |
XM_005258477.3 | 1704 | Intron | XP_005258534.2 |