Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCACCTTCTTCACAAAATGCTC[C/T]ATGGCAATTTCTGTAGTGGGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609475 | ||||||||||||||||||||
Literature Links: |
AKAP8L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291478.1 | 1521 | Missense Mutation | NP_001278407.1 | |||
NM_014371.3 | 1521 | Missense Mutation | NP_055186.3 | |||
XM_005259854.3 | 1521 | Missense Mutation | XP_005259911.2 | |||
XM_011527916.2 | 1521 | Missense Mutation | XP_011526218.1 |