Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACGGTATGGCTGGGTCCCATCA[C/T]CCCCTTCATCGTTTTATGCCACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611485 | ||||||||||||||||||||
Literature Links: |
CYP4F12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP4F12 - cytochrome P450 family 4 subfamily F member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023944.3 | 348 | Intron | NP_076433.3 | |||
XM_006722850.3 | 348 | Intron | XP_006722913.1 | |||
XM_011528202.2 | 348 | Intron | XP_011526504.1 | |||
XM_011528203.2 | 348 | Intron | XP_011526505.1 | |||
XM_011528204.2 | 348 | Intron | XP_011526506.1 | |||
XM_011528205.2 | 348 | Intron | XP_011526507.1 | |||
XM_011528207.2 | 348 | Intron | XP_011526509.1 | |||
XM_011528208.2 | 348 | Intron | XP_011526510.1 | |||
XM_017027172.1 | 348 | Missense Mutation | ACC,ATC | T,I 96 | XP_016882661.1 |