Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTGAAAGGACTTATGAGTGCAGA[C/G]AATGTGGAAAATCCTTTATGTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF548 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF548 - zinc finger protein 548 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172773.1 | 1055 | Missense Mutation | CAA,GAA | Q,E 269 | NP_001166244.1 | |
NM_152909.3 | 1055 | Missense Mutation | CAA,GAA | Q,E 257 | NP_690873.2 | |
XM_017026316.1 | 1055 | Missense Mutation | CAA,GAA | Q,E 260 | XP_016881805.1 |