Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAGCTGCCTTCTGGGCTGGGCTC[A/G]GGCGAGGGCAGCAGGTGGGCAAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614585 MIM: 609950 MIM: 611639 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FDX1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FDX1L - ferredoxin 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAVER1 - ribonucleoprotein, PTB binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133452.2 | 2255 | Silent Mutation | CCC,CCT | P,P 725 | NP_597709.2 | |
XM_011527671.1 | 2255 | Silent Mutation | CCC,CCT | P,P 700 | XP_011525973.1 |
ZGLP1 - zinc finger, GATA-like protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |