Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCATCGCCCACAAGCTGCAGAAT[A/G]AGCCCTTCGCTGGTGGAGAGGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612928 | ||||||||||||||||||||
Literature Links: |
ISOC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISOC2 - isochorismatase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136201.1 | 666 | Silent Mutation | CTC,CTT | L,L 164 | NP_001129673.1 | |
NM_001136202.1 | 666 | Silent Mutation | CTC,CTT | L,L 94 | NP_001129674.1 | |
NM_024710.2 | 666 | Silent Mutation | CTC,CTT | L,L 180 | NP_078986.1 |