Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTTCGTAGGTTTTGACGCAGAAG[A/G]TCAGGCGGGGGTTGGGGCTCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYPD3 - LY6/PLAUR domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHLDB3 - pleckstrin homology like domain family B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198850.3 | 2239 | Missense Mutation | ACC,ATC | T,I 603 | NP_942147.3 | |
XM_005259169.3 | 2239 | Missense Mutation | ACC,ATC | T,I 598 | XP_005259226.1 | |
XM_005259170.3 | 2239 | Missense Mutation | ACC,ATC | T,I 564 | XP_005259227.1 | |
XM_005259171.3 | 2239 | Intron | XP_005259228.1 | |||
XM_011527236.2 | 2239 | Missense Mutation | ACC,ATC | T,I 598 | XP_011525538.1 |