Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACGTGGCCATGAACCGCCGGG[C/G]CCGCCTCTACGGGTCGGTGAGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605831 MIM: 601778 MIM: 615177 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF22 - fibroblast growth factor 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300812.1 | 333 | Missense Mutation | GCC,GGC | A,G 101 | NP_001287741.1 | |
NM_020637.1 | 333 | Missense Mutation | GCC,GGC | A,G 101 | NP_065688.1 |
POLRMT - RNA polymerase mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF126 - ring finger protein 126 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |