Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACAGGCCCAGGGCTGGGGCACA[C/T]TGTGCTTCTGCAAGGCTCGCCGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGL3 - ral guanine nucleotide dissociation stimulator like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035223.3 | 2037 | Missense Mutation | AAT,AGT | N,S 652 | NP_001030300.3 | |
NM_001161616.2 | 2037 | Missense Mutation | AAT,AGT | N,S 658 | NP_001155088.2 |