Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCGCGGTGCACCAGGTGATGAT[C/T]GTGCAGGTAGCGCACGGCGCCGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614997 MIM: 609435 MIM: 610712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATAD2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATAD2A - GATA zinc finger domain containing 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA13 - NADH:ubiquinone oxidoreductase subunit A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSSK6 - testis specific serine kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032037.3 | 615 | Missense Mutation | NP_114426.1 |