Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGCTGGCGGGCTCTGAGCCGT[C/T]GTGCTGGGCGCAGGGCTCCTGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611998 MIM: 606211 | ||||||||||||||||||||
Literature Links: |
ANKRD24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD24 - ankyrin repeat domain 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CREB3L3 - cAMP responsive element binding protein 3 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRT6 - sirtuin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193285.2 | 852 | Missense Mutation | AAC,GAC | N,D 297 | NP_001180214.1 | |
NM_001321058.1 | 852 | Missense Mutation | AAC,GAC | N,D 252 | NP_001307987.1 | |
NM_001321059.1 | 852 | Missense Mutation | AAC,GAC | N,D 263 | NP_001307988.1 | |
NM_001321060.1 | 852 | UTR 3 | NP_001307989.1 | |||
NM_001321061.1 | 852 | Missense Mutation | AAC,GAC | N,D 225 | NP_001307990.1 | |
NM_001321062.1 | 852 | Missense Mutation | AAC,GAC | N,D 189 | NP_001307991.1 | |
NM_001321063.1 | 852 | UTR 3 | NP_001307992.1 | |||
NM_001321064.1 | 852 | UTR 3 | NP_001307993.1 | |||
NM_016539.3 | 852 | Missense Mutation | AAC,GAC | N,D 324 | NP_057623.2 |