Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTGAGCCATAGCCACTGCTTT[C/T]GAACACATGAAAGGCCTCACACTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF440 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF440 - zinc finger protein 440 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF491 - zinc finger protein 491 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152356.3 | 497 | Nonsense Mutation | CGA,TGA | R,* 91 | NP_689569.2 | |
XM_005259730.3 | 497 | Nonsense Mutation | CGA,TGA | R,* 91 | XP_005259787.1 |