Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCGCAGTGACCAGGCCAGCATCA[A/G]CCTGCGACTGACCTCCCCACTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF324B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF324B - zinc finger protein 324B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207395.2 | 607 | Missense Mutation | AAC,AGC | N,S 159 | NP_997278.2 | |
XM_005258915.4 | 607 | Missense Mutation | AAC,AGC | N,S 159 | XP_005258972.1 | |
XM_005258918.4 | 607 | Missense Mutation | AAC,AGC | N,S 159 | XP_005258975.1 |