Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGACCTCAAACAGAAGTCCCT[G/T]TACAGGGAAGTGATGCTGGAGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605467 | ||||||||||||||||||||
Literature Links: |
ZNF274 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF274 - zinc finger protein 274 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278734.1 | 613 | UTR 5 | NP_001265663.1 | |||
NM_016324.3 | 613 | Intron | NP_057408.2 | |||
NM_016325.3 | 613 | Silent Mutation | CTG,CTT | L,L 38 | NP_057409.1 | |
NM_133502.2 | 613 | Silent Mutation | CTG,CTT | L,L 38 | NP_598009.1 | |
XM_011526327.1 | 613 | Intron | XP_011524629.1 | |||
XM_011526328.2 | 613 | Intron | XP_011524630.1 | |||
XM_017026174.1 | 613 | Intron | XP_016881663.1 | |||
XM_017026175.1 | 613 | Silent Mutation | CTG,CTT | L,L 38 | XP_016881664.1 |