Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCTCAAGGAGCAGCTCAAGCTC[C/T]GCCTGGCCGCCGAGGGCTTCGGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180901 | ||||||||||||||||||||
Literature Links: |
RYR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RYR1 - ryanodine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000540.2 | 183 | Missense Mutation | CGC,TGC | R,C 35 | NP_000531.2 | |
NM_001042723.1 | 183 | Missense Mutation | CGC,TGC | R,C 35 | NP_001036188.1 | |
XM_006723317.2 | 183 | Missense Mutation | CGC,TGC | R,C 35 | XP_006723380.1 | |
XM_006723319.2 | 183 | Missense Mutation | CGC,TGC | R,C 35 | XP_006723382.1 | |
XM_011527205.2 | 183 | Missense Mutation | CGC,TGC | R,C 35 | XP_011525507.1 |