Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGATGAAACTCAATTTAAGGCC[A/G]GTGGGTCAGTTTCTCAGCAGGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF555 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF555 - zinc finger protein 555 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172775.1 | 295 | Missense Mutation | AGT,GGT | S,G 52 | NP_001166246.1 | |
NM_152791.4 | 295 | Missense Mutation | AGT,GGT | S,G 52 | NP_690004.4 | |
XM_011527716.2 | 295 | Missense Mutation | AGT,GGT | S,G 54 | XP_011526018.1 | |
XM_017026375.1 | 295 | Missense Mutation | AGT,GGT | S,G 54 | XP_016881864.1 |