Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCCAGCTACTGCGAAGAAAGC[A/C]GGGGCAACAACCAGTGGGTCCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604948 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GADD45B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GADD45B - growth arrest and DNA damage inducible beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015675.3 | 670 | Silent Mutation | AGG,CGG | R,R 146 | NP_056490.2 | |
XM_017026822.1 | 670 | UTR 3 | XP_016882311.1 |