Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGATGTTTAGTGAGGCCTGAGCG[A/G]CAAATAAAAGATTTCCCACACTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF616 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF616 - zinc finger protein 616 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178523.4 | 2551 | Silent Mutation | TGC,TGT | C,C 756 | NP_848618.2 | |
XM_017027438.1 | 2551 | Silent Mutation | TGC,TGT | C,C 787 | XP_016882927.1 |