Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAAAGCCGCTGAGGACAAGTGC[A/G]AGCAGGTGAGGTGCCCTCCGCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600317 | ||||||||||||||||||||
Literature Links: |
TPM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPM4 - tropomyosin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145160.1 | 34 | Missense Mutation | AAG,GAG | K,E 37 | NP_001138632.1 | |
NM_003290.2 | 34 | Intron | NP_003281.1 | |||
XM_005260042.3 | 34 | Intron | XP_005260099.1 | |||
XM_006722865.2 | 34 | Missense Mutation | AAG,GAG | K,E 37 | XP_006722928.1 | |
XM_011528231.2 | 34 | Intron | XP_011526533.1 | |||
XM_011528232.1 | 34 | Intron | XP_011526534.1 | |||
XM_017027188.1 | 34 | UTR 5 | XP_016882677.1 | |||
XM_017027189.1 | 34 | Intron | XP_016882678.1 |