Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTATGGGTACACAACGCCACTGAC[G/T]GATGCGGGCAAGGCCTTCTCCATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613452 MIM: 603939 | ||||||||||||||||||||
Literature Links: |
CATSPERG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CATSPERG - cation channel sperm associated auxiliary subunit gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK6 - potassium two pore domain channel subfamily K member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004823.1 | 1020 | Silent Mutation | ACG,ACT | T,T 116 | NP_004814.1 | |
XM_011527526.1 | 1020 | UTR 5 | XP_011525828.1 | |||
XM_011527527.1 | 1020 | Intron | XP_011525829.1 | |||
XM_017027506.1 | 1020 | Intron | XP_016882995.1 |