Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGGCAGCCTCCCCCAGGAGGAG[A/G]CTGGAGAGAGGCCAGAGTCCGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PBX4 - PBX homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025245.2 | 968 | Missense Mutation | CCT,TCT | P,S 334 | NP_079521.1 | |
XM_006722911.3 | 968 | Missense Mutation | CCT,TCT | P,S 248 | XP_006722974.1 | |
XM_011528320.2 | 968 | Missense Mutation | CCT,TCT | P,S 351 | XP_011526622.1 | |
XM_011528322.2 | 968 | Missense Mutation | CCT,TCT | P,S 248 | XP_011526624.1 | |
XM_017027326.1 | 968 | Missense Mutation | CCT,TCT | P,S 317 | XP_016882815.1 | |
XM_017027327.1 | 968 | Missense Mutation | CCT,TCT | P,S 303 | XP_016882816.1 | |
XM_017027328.1 | 968 | Intron | XP_016882817.1 | |||
XM_017027329.1 | 968 | Missense Mutation | CCT,TCT | P,S 169 | XP_016882818.1 | |
XM_017027330.1 | 968 | Intron | XP_016882819.1 |