Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGCATTCTCAACAGCAACAGC[A/G]TGGCCTGCAGCTTTGCCGTGGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 131235 MIM: 608373 | ||||||||||||||||||||
Literature Links: |
KDELR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDELR1 - KDEL endoplasmic reticulum protein retention receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGR4 - synaptogyrin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012451.3 | 675 | Missense Mutation | ATG,GTG | M,V 64 | NP_036583.2 | |
XM_005258693.2 | 675 | Missense Mutation | ATG,GTG | M,V 64 | XP_005258750.1 | |
XM_005258694.2 | 675 | Missense Mutation | ATG,GTG | M,V 64 | XP_005258751.1 | |
XM_011526686.2 | 675 | Missense Mutation | ATG,GTG | M,V 15 | XP_011524988.1 | |
XM_017026541.1 | 675 | Missense Mutation | ATG,GTG | M,V 15 | XP_016882030.1 |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |