Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGCCTCTGTCCCCGCAGGAGGT[A/G]TTGCCCATGGCCCCCGGGGCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL4 - mitochondrial ribosomal protein L4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015956.2 | 605 | Missense Mutation | ATT,GTT | I,V 151 | NP_057040.2 | |
NM_146387.1 | 605 | Missense Mutation | ATT,GTT | I,V 151 | NP_666499.1 | |
NM_146388.1 | 605 | Missense Mutation | ATT,GTT | I,V 151 | NP_666500.1 | |
XM_011528045.2 | 605 | Missense Mutation | ATT,GTT | I,V 151 | XP_011526347.1 |