Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCCCGGTCGGCCGCCGCTCCCC[A/G]CCTAGTCCACGGAGCTCAATGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613419 | ||||||||||||||||||||
Literature Links: |
ZNF551 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF551 - zinc finger protein 551 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270938.1 | 225 | Intron | NP_001257867.1 | |||
NM_138347.4 | 225 | Silent Mutation | CCA,CCG | P,P 10 | NP_612356.2 |
ZSCAN4 - zinc finger and SCAN domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |