Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAAGGGGAAGCCGCAGCTCCA[A/G]GAGGTCACCCAGCAGGGCCAGCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609744 | ||||||||||||||||||||
Literature Links: |
CADM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CADM4 - cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRRM5 - serine/arginine repetitive matrix 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145641.1 | 419 | Missense Mutation | AAG,AGG | K,R 133 | NP_001139113.1 |
ZNF428 - zinc finger protein 428 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182498.3 | 419 | Intron | NP_872304.2 |