Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTCGGGTTCCCCCCGGGTCGCC[C/T]GCAGCTCCCGGTCCAGCTCCGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604076 MIM: 601856 | ||||||||||||||||||||
Literature Links: |
ZNF134 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF134 - zinc finger protein 134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF211 - zinc finger protein 211 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001265597.2 | 206 | Missense Mutation | CCG,CTG | P,L 9 | NP_001252526.1 | |
NM_001265598.2 | 206 | Missense Mutation | CCG,CTG | P,L 9 | NP_001252527.1 | |
NM_001265599.2 | 206 | Intron | NP_001252528.1 | |||
NM_001265600.2 | 206 | Intron | NP_001252529.1 | |||
NM_001322306.1 | 206 | Missense Mutation | CCG,CTG | P,L 9 | NP_001309235.1 | |
NM_006385.4 | 206 | Missense Mutation | CCG,CTG | P,L 9 | NP_006376.2 | |
NM_198855.3 | 206 | Missense Mutation | CCG,CTG | P,L 9 | NP_942152.1 |