Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGTTTTGAGTCAGGACCCATC[A/T]TGCTGGAAAGTGTGTATGTCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRE3 - adhesion G protein-coupled receptor E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289158.1 | 1334 | Missense Mutation | AAG,ATG | K,M 579 | NP_001276087.1 | |
NM_001289159.1 | 1334 | Missense Mutation | AAG,ATG | K,M 505 | NP_001276088.1 | |
NM_032571.4 | 1334 | Missense Mutation | AAG,ATG | K,M 631 | NP_115960.2 | |
XM_011528374.2 | 1334 | Missense Mutation | AAG,ATG | K,M 614 | XP_011526676.1 | |
XM_017027383.1 | 1334 | Missense Mutation | AAG,ATG | K,M 416 | XP_016882872.1 |
SNORA104 - small nucleolar RNA, H/ACA box 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |