Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCAGGAGATACCTTCCCGGATG[C/T]AGATGCTGATGAAGACAGTCTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605350 MIM: 606855 | ||||||||||||||||||||
Literature Links: |
IL27RA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL27RA - interleukin 27 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RLN3 - relaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311197.1 | 378 | Silent Mutation | TGC,TGT | C,C 107 | NP_001298126.1 | |
NM_080864.3 | 378 | Missense Mutation | GCA,GTA | A,V 70 | NP_543140.1 |