Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTCATGGCCCTCCTGCAGCAC[A/G]TGGCCCTGCAGTTTGAGCCAGGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601855 MIM: 112205 | ||||||||||||||||||||
Literature Links: |
ARHGEF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF1 - Rho guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004706.3 | 653 | Missense Mutation | ATG,GTG | M,V 66 | NP_004697.2 | |
NM_198977.1 | 653 | Missense Mutation | ATG,GTG | M,V 66 | NP_945328.1 | |
NM_199002.1 | 653 | Missense Mutation | ATG,GTG | M,V 81 | NP_945353.1 | |
XM_005259386.4 | 653 | Missense Mutation | ATG,GTG | M,V 81 | XP_005259443.1 | |
XM_005259390.4 | 653 | Missense Mutation | ATG,GTG | M,V 81 | XP_005259447.1 | |
XM_011527468.2 | 653 | Missense Mutation | ATG,GTG | M,V 81 | XP_011525770.1 |
CD79A - CD79a molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |