Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCCCTGGATGCCCACCTCCACC[A/G]GTGCATATTTGACCGGAGGTTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP33 - Rho GTPase activating protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172630.1 | 1875 | UTR 5 | NP_001166101.1 | |||
NM_052948.3 | 1875 | Missense Mutation | CAG,CGG | Q,R 112 | NP_443180.2 | |
XM_005258488.1 | 1875 | Missense Mutation | CAG,CGG | Q,R 112 | XP_005258545.1 | |
XM_006722999.1 | 1875 | Missense Mutation | CAG,CGG | Q,R 112 | XP_006723062.1 | |
XM_006723003.2 | 1875 | Intron | XP_006723066.1 | |||
XM_011526416.1 | 1875 | UTR 5 | XP_011524718.1 | |||
XM_011526417.1 | 1875 | UTR 5 | XP_011524719.1 | |||
XM_011526418.1 | 1875 | UTR 5 | XP_011524720.1 | |||
XM_011526419.1 | 1875 | UTR 5 | XP_011524721.1 | |||
XM_011526420.1 | 1875 | UTR 5 | XP_011524722.1 | |||
XM_011526421.2 | 1875 | Missense Mutation | CAG,CGG | Q,R 149 | XP_011524723.1 | |
XM_017026238.1 | 1875 | Missense Mutation | CAG,CGG | Q,R 149 | XP_016881727.1 |
PROSER3 - proline and serine rich 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |