Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTCAGGGTCTCACTTGAAGTAG[A/C]GATTGGGTTTGTAGTGCAGTTTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601480 MIM: 606466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO1F - myosin IF | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAM1 - PML-RARA regulated adaptor molecule 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF414 - zinc finger protein 414 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146175.1 | 639 | Missense Mutation | CGC,CTC | R,L 174 | NP_001139647.1 | |
NM_032370.2 | 639 | Missense Mutation | CGC,CTC | R,L 174 | NP_115746.2 |