Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGTACAAGGCGGTCTCCGTGCTC[C/T]GCTGCGCCGCATGCTTCTTCACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604840 | ||||||||||||||||||||
Literature Links: |
FKBP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKBP8 - FK506 binding protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308373.1 | 1267 | Missense Mutation | CAG,CGG | Q,R 357 | NP_001295302.1 | |
NM_012181.4 | 1267 | Missense Mutation | CAG,CGG | Q,R 358 | NP_036313.3 | |
XM_011527863.1 | 1267 | Missense Mutation | CAG,CGG | Q,R 358 | XP_011526165.1 | |
XM_011527864.1 | 1267 | Silent Mutation | GCA,GCG | A,A 364 | XP_011526166.1 |