Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCTCCTCTCTCGTTTCAGCGA[A/G]CACCTGTGAGGAGTGGGCGGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608792 MIM: 605535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIPC3 - GIPC PDZ domain containing family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMG20B - high mobility group 20B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006339.2 | 1013 | Silent Mutation | GAA,GAG | E,E 315 | NP_006330.2 | |
XM_017026144.1 | 1013 | Silent Mutation | GAA,GAG | E,E 314 | XP_016881633.1 |