Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCTCACCACGCACGTTCCGCC[A/G]CCGGATCGCCTAGGAAGGAGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPK3 - death associated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001348.2 | 1135 | Missense Mutation | CGG,TGG | R,W 280 | NP_001339.1 | |
XM_005259508.1 | 1135 | Missense Mutation | CGG,TGG | R,W 280 | XP_005259565.1 |
MIR637 - microRNA 637 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |