Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGGGCCAGGATGTCAAGGGGC[A/G]GAGTTCCGGGTTCCACAGCTGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614566 MIM: 191044 MIM: 191041 | ||||||||||||||||||||
Literature Links: |
DNAAF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAAF3 - dynein axonemal assembly factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256714.1 | 1636 | Missense Mutation | CCG,CTG | P,L 541 | NP_001243643.1 | |
NM_001256715.1 | 1636 | Missense Mutation | CCG,CTG | P,L 474 | NP_001243644.1 | |
NM_001256716.1 | 1636 | Missense Mutation | CCG,CTG | P,L 420 | NP_001243645.1 | |
NM_178837.4 | 1636 | Missense Mutation | CCG,CTG | P,L 521 | NP_849159.2 |
TNNI3 - troponin I3, cardiac type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNT1 - troponin T1, slow skeletal type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |