Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGTTACATTGAAACGGCTTCTC[C/T]CCAGTATGAATTCTCTTATGATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF836 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF836 - zinc finger protein 836 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102657.1 | 2311 | Silent Mutation | GGA,GGG | G,G 634 | NP_001096127.1 | |
XM_011526558.2 | 2311 | Silent Mutation | GGA,GGG | G,G 634 | XP_011524860.1 | |
XM_011526559.2 | 2311 | Silent Mutation | GGA,GGG | G,G 634 | XP_011524861.1 | |
XM_017026389.1 | 2311 | Intron | XP_016881878.1 |