Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCAAGCCAGGAGGCCGGCTCG[A/G]GCCAGGCTGCCAAAGAGGATGAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606048 MIM: 603631 MIM: 608754 | ||||||||||||||||||||
Literature Links: |
MBOAT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBOAT7 - membrane bound O-acyltransferase domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS9 - ribosomal protein S9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSEN34 - tRNA splicing endonuclease subunit 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077446.3 | 527 | Missense Mutation | AGC,GGC | S,G 142 | NP_001070914.1 | |
NM_001282332.1 | 527 | Missense Mutation | AGC,GGC | S,G 142 | NP_001269261.1 | |
NM_001282333.1 | 527 | Missense Mutation | AGC,GGC | S,G 145 | NP_001269262.1 | |
NM_024075.4 | 527 | Missense Mutation | AGC,GGC | S,G 142 | NP_076980.2 | |
XM_005278290.4 | 527 | Missense Mutation | AGC,GGC | S,G 145 | XP_005278347.1 | |
XM_011527294.2 | 527 | Missense Mutation | AGC,GGC | S,G 142 | XP_011525596.1 | |
XM_011527295.1 | 527 | Missense Mutation | AGC,GGC | S,G 142 | XP_011525597.1 |