Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCTCCTGTTTGACCGGCAGGAC[A/G]GCATCCTGAGACACGTGGAGCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611998 | ||||||||||||||||||||
Literature Links: |
CREB3L3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CREB3L3 - cAMP responsive element binding protein 3 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271995.1 | 250 | Missense Mutation | AGC,GGC | S,G 35 | NP_001258924.1 | |
NM_001271996.1 | 250 | Missense Mutation | AGC,GGC | S,G 35 | NP_001258925.1 | |
NM_001271997.1 | 250 | Missense Mutation | AGC,GGC | S,G 35 | NP_001258926.1 | |
NM_032607.2 | 250 | Missense Mutation | AGC,GGC | S,G 35 | NP_115996.1 |