Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCTGTGGTGCGGGGTTCCAGC[A/C]AGCGGGCGTTGACAGTGGCCAGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604350 MIM: 613771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC159 - coiled-coil domain containing 159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3D - RAB3D, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM205 - transmembrane protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145416.1 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_001138888.1 | |
NM_001321112.1 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_001308041.1 | |
NM_001321113.1 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_001308042.1 | |
NM_001321114.1 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_001308043.1 | |
NM_033408.3 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_212133.1 | |
NM_198536.2 | 312 | Missense Mutation | TGG,TTG | W,L 105 | NP_940938.1 | |
XM_005259899.2 | 312 | Missense Mutation | TGG,TTG | W,L 105 | XP_005259956.1 | |
XM_017026769.1 | 312 | Silent Mutation | CTG,CTT | L,L 98 | XP_016882258.1 |