Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTGGTTCTCATAGTCAACAGCT[G/T]GGCCATCTCCAATGGGGGTCACGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609828 MIM: 607881 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSD1 - fibronectin type III and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAP2 - signal transducing adaptor family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013841.1 | 1050 | Missense Mutation | CAA,CCA | Q,P 318 | NP_001013863.1 | |
NM_017720.2 | 1050 | Missense Mutation | CAA,CCA | Q,P 318 | NP_060190.2 | |
XM_011528123.1 | 1050 | Missense Mutation | CAA,CCA | Q,P 318 | XP_011526425.1 |