Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTTGTGCCCTTCCGCTGCCTG[C/T]GTGAGTCCCAGGCGGGGAGAGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104775 MIM: 607762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APLP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APLP1 - amyloid beta precursor like protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024807.2 | 616 | Missense Mutation | CCT,TCT | P,S 142 | NP_001019978.1 | |
NM_005166.4 | 616 | Missense Mutation | CCT,TCT | P,S 142 | NP_005157.1 | |
XM_017026737.1 | 616 | Missense Mutation | CCT,TCT | P,S 142 | XP_016882226.1 | |
XM_017026738.1 | 616 | Missense Mutation | CCT,TCT | P,S 142 | XP_016882227.1 |
KIRREL2 - kin of IRRE like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |