Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCGCTGTCGGGGCCCGCGTCGG[G/T]GAAGGCGCCAGTGCGCAGCGTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEX3D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEX3D - mex-3 RNA binding family member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174118.1 | 1036 | Missense Mutation | ACC,CCC | T,P 346 | NP_001167589.1 | |
NM_203304.3 | 1036 | Missense Mutation | ACC,CCC | T,P 346 | NP_976049.3 | |
XM_017026811.1 | 1036 | Missense Mutation | ACC,CCC | T,P 112 | XP_016882300.1 |