Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGATTCCTCAAACAGGCCTGAG[C/T]CTCTGCAGTTTCTCGCTTGTGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605639 | ||||||||||||||||||||
Literature Links: |
SIGLEC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIGLEC8 - sialic acid binding Ig like lectin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014442.2 | 1475 | Missense Mutation | ACT,GCT | T,A 483 | NP_055257.2 | |
XM_011526734.2 | 1475 | Missense Mutation | ACT,GCT | T,A 472 | XP_011525036.1 | |
XM_011526735.2 | 1475 | Missense Mutation | ACT,GCT | T,A 390 | XP_011525037.1 |