Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCCGTGTGGAAGGCGCTCTTCG[G/T]CAAGGAGGCGGACAAGCTGGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151445 MIM: 609565 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCER2 - Fc fragment of IgE receptor II | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCEMP1 - mast cell expressed membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC5 - trafficking protein particle complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042461.2 | 366 | Missense Mutation | GGC,GTC | G,V 99 | NP_001035926.1 | |
NM_001042462.1 | 366 | Missense Mutation | GGC,GTC | G,V 99 | NP_001035927.1 | |
NM_174894.2 | 366 | Missense Mutation | GGC,GTC | G,V 99 | NP_777554.1 |